Skip to Content
Merck
All Photos(1)

Key Documents

181609

Sigma-Aldrich

Polycaprolactone

average Mw ~65,000, average Mn ~42,500, pellets

Synonym(s):

2-Oxepanone homopolymer, 6-Caprolactone polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O2)n
UNSPSC Code:
12162002

form

pellets

melt index

1.9 g/10 min (80°C/0.3 Mpa)

mol wt

average Mn ~42,500
average Mw ~65,000

impact strength

82 J/m (Izod, ASTM D 256-73A, notched)

hardness

55 (Shore D, ASTM D 2240-75)

mp

60 °C (lit.)

elongation

2 in/min - 600-1000% (ultimate)

Looking for similar products? Visit Product Comparison Guide

Application

Extrusion aid, die lubricant, mold release, pigment and filler dispersion aid and polyester segments in urethanes and block polyesters.

Features and Benefits

Biodegradable polymer
Non-toxic, biodegradable in soil, broad miscibility, mechanical compatibility with many polymers and good adhesion to a broad spectrum of substrates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kalaipriya Madhaiyan et al.
International journal of pharmaceutics, 444(1-2), 70-76 (2013-02-02)
Biocompatible PCL polymer nanofiber mediated sustained release of hydrophilic drug and applicability as transdermal delivery system is attempted. This new attempt to investigate water soluble vitamin delivery with hydrophobic polymer nanofiber sustained the release of the vitamin and the method
Qingchun Zhang et al.
Materials science & engineering. C, Materials for biological applications, 33(4), 2094-2103 (2013-03-19)
The physical properties of tissue engineering scaffolds such as microstructures play important roles in controlling cellular behaviors and neotissue formation. Among them, the pore size stands out as a key determinant factor. In the present study, we aimed to fabricate
P R Sreerekha et al.
Journal of biomedical nanotechnology, 9(5), 790-800 (2013-06-28)
Fabricating scaffolds mimicking the native extracellular matrix (ECM) in both structure and function is a key challenge in the field of tissue engineering. Previously we have demonstrated a novel electrospinnig method for the fabrication of fibrin nanofibers using Poly(vinyl alcohol)
Trung Dung Dao et al.
Journal of nanoscience and nanotechnology, 12(11), 8420-8430 (2013-02-21)
A functionalized graphene sheet (FGS), which was prepared by the thermal reduction of graphite oxide, was modified by oxidation with H2O2. Elemental analysis, X-ray photoelectron spectroscopy, and Fourier transform infrared (FTIR) spectroscopy showed that additional oxygen functional groups, either doubly
Richard A Thibault et al.
Journal of biomedical materials research. Part A, 101(5), 1225-1236 (2013-03-19)
Devitalization and demineralization processing of biodegradable polymer and extracellular matrix (ECM) hybrid constructs was explored for the effect on the retention of ECM components and construct osteogenicity. Hybrid constructs were generated by seeding osteogenically predifferentiated rat mesenchymal stem cells (MSCs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service