Skip to Content
Merck
All Photos(1)

Key Documents

180858

Sigma-Aldrich

1,1′-Diheptyl-4,4′-bipyridinium dibromide

97%

Synonym(s):

N ,N ′-Diheptyl-4,4′-dipyridylium dibromide, DHPB, Diheptylviologen bromide, Diheptylviologen dibromide, Heptylviologen dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38Br2N2
CAS Number:
Molecular Weight:
514.38
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

crystals

mp

285 °C (dec.) (lit.)

SMILES string

[Br-].[Br-].CCCCCCC[n+]1ccc(cc1)-c2cc[n+](CCCCCCC)cc2

InChI

1S/C24H38N2.2BrH/c1-3-5-7-9-11-17-25-19-13-23(14-20-25)24-15-21-26(22-16-24)18-12-10-8-6-4-2;;/h13-16,19-22H,3-12,17-18H2,1-2H3;2*1H/q+2;;/p-2

InChI key

VRXAJMCFEOESJO-UHFFFAOYSA-L

General description

1,1′-Diheptyl-4,4′-bipyridinium dibromide (DHBP) is a heptyl viologen that can be used as an electrochromic (EC) material. It forms a highly efficient quenching system for conjugated polyelectrolyte for the development of organic electronics. It also acts as a redox active electrolyte for electrochemical devices (ECDs).

Application

DHBP is an EC material which can be used in the fabrication of electronic devices such as organic light emitting diodes (OLED), supercapacitors, and liquid crystal displays (LCDs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of ferrocyanide on the performance of heptyl viologen-based electrochromic display devices.
Monk PMS.
Journal of Electroanalytical Chemistry, 432(1-2), 175-179 (1997)
Electrochemically controllable emission and coloration by using europium (III) complex and viologen derivatives.
Nakamura K, et al.
Chemical Communications (Cambridge, England), 47(36), 10064-10066 (2011)
Achieving low-energy driven viologens-based electrochromic devices utilizing polymeric ionic liquids.
Lu H, et al.
ACS Applied Materials & Interfaces, 8(44), 30351-30361 (2016)
Ink-Jet Printing of Electron Donor/Acceptor Blends: Towards Bulk Heterojunction Solar Cells
Marin, V., Holder, E., Wienk, M. M., Tekin, E., Kozodaev, D., & Schubert, U. S.
Macromolecular Rapid Communications, 26(4), 319-324 (2005)
Efficient charge storage in dual-redox electrochemical capacitors through reversible counterion-induced solid complexation.
Evanko B, et al.
Journal of the American Chemical Society, 138(30), 9373-9376 (2016)

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service