Skip to Content
Merck
All Photos(1)

Key Documents

178241

Sigma-Aldrich

Isobutyl 1,2-dihydro-2-isobutoxy-1-quinolinecarboxylate

99%

Synonym(s):

1-Isobutoxycarbonyl-2-isobutoxy-1,2-dihydroquinoline, 2-Isobutoxy-1-isobutoxycarbonyl-1,2-dihydroquinoline, IIDQ

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$595.00

$595.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$595.00

About This Item

Empirical Formula (Hill Notation):
C18H25NO3
CAS Number:
Molecular Weight:
303.40
Beilstein:
1543065
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$595.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

99%

reaction suitability

reaction type: Coupling Reactions

refractive index

n20/D 1.523 (lit.)

bp

140-142 °C/0.2 mmHg (lit.)

density

1.022 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

functional group

ether

SMILES string

CC(C)COC1C=Cc2ccccc2N1C(=O)OCC(C)C

InChI

1S/C18H25NO3/c1-13(2)11-21-17-10-9-15-7-5-6-8-16(15)19(17)18(20)22-12-14(3)4/h5-10,13-14,17H,11-12H2,1-4H3

InChI key

LPBHYOYZZIFCQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manisha Sathe et al.
Bioorganic & medicinal chemistry letters, 20(5), 1792-1795 (2010-02-05)
Polystyrene-supported 2-isobutoxy-1-isobutoxycarbonyl-1,2-dihydroquinoline (PS-IIDQ), a polymer-supported covalent coupling reagent, was successfully employed for the first time in the bioconjugation of an example hapten (phytanic acid derivative) to a carrier protein (bovine serum albumin (BSA)) within the context of immunogen preparation for
Effect of hydrophobic carboxyl reagents on the proton flux through coupling factor CF0 in thylakoid membrane.
Y Ho et al.
Biochemistry, 19(12), 2650-2655 (1980-06-10)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service