Skip to Content
Merck
All Photos(1)

Key Documents

555215

Sigma-Aldrich

ScavengePore phenethyl diethylamine

macroporous, 40-70 mesh, extent of labeling: 0.7-1.5 mmol/g loading

Synonym(s):

MP diethylamine resin, Phenethyl diamine, polymer-bound

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$339.00

$339.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$339.00

About This Item

MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$339.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

product line

ScavengePore

form

macroporous

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

0.7-1.5 mmol/g loading

particle size

40-70 mesh

functional group

amine

storage temp.

2-8°C

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.CCN(CC)CCc3ccccc3

InChI

1S/C12H19N.C10H10.C8H8/c1-3-13(4-2)11-10-12-8-6-5-7-9-12;1-3-9-5-7-10(4-2)8-6-9;1-2-8-6-4-3-5-7-8/h5-9H,3-4,10-11H2,1-2H3;3-8H,1-2H2;2-7H,1H2

InChI key

MFCQPWPGLCCHOO-UHFFFAOYSA-N

Legal Information

ScavengePore is a trademark of Rapp Polymere GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service