Skip to Content
Merck
All Photos(1)

Documents

177857

Sigma-Aldrich

1-Iodoheptane

98%

Synonym(s):

Heptyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6I
CAS Number:
Molecular Weight:
226.10
Beilstein:
1697161
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.490 (lit.)

bp

204 °C (lit.)

mp

−48 °C (lit.)

solubility

benzene: miscible(lit.)
chloroform: miscible(lit.)

density

1.379 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCI

InChI

1S/C7H15I/c1-2-3-4-5-6-7-8/h2-7H2,1H3

InChI key

LMHCYRULPLGEEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1-Iodoheptane was used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barada Prasanna Dash et al.
Organic letters, 10(11), 2247-2250 (2008-05-10)
Symmetrical star-shaped molecules with carborane clusters on the periphery have been synthesized in good yields via silicon tetrachloride mediated cyclotrimerization reactions of 9-benzyl derivatives of carboranes with acetyl group substitution on the benzene ring. Facile functionalization of these symmetrical core

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service