Skip to Content
Merck
All Photos(1)

Key Documents

173681

Sigma-Aldrich

Methyl 4-methoxysalicylate

98%

Synonym(s):

Methyl 2-hydroxy-4-methoxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(OH)CO2CH3
CAS Number:
Molecular Weight:
182.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

50-53 °C (lit.)

SMILES string

COC(=O)c1ccc(OC)cc1O

InChI

1S/C9H10O4/c1-12-6-3-4-7(8(10)5-6)9(11)13-2/h3-5,10H,1-2H3

InChI key

ZICRWXFGZCVTBZ-UHFFFAOYSA-N

Application

Methyl 4-methoxysalicylate (methyl 2-hydroxy-4-methoxybenzoate) was used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjing Wang et al.
Organic & biomolecular chemistry, 11(39), 6686-6690 (2013-08-30)
The first enantioselective total synthesis of (+)-coriandrone A and B, two bioactive natural products, has been achieved in 10 steps and 11 steps starting from commercially available methyl 2-hydroxy-4-methoxybenzoate. Key reactions include a Claison rearrangement, a Shi-type epoxidation-cyclization sequence and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service