Skip to Content
Merck
All Photos(1)

Key Documents

122068

Sigma-Aldrich

trans-4-Methoxy-3-buten-2-one

technical grade, 90%

Synonym(s):

(E)-4-Methoxy-3-buten-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$127.00
50 G
$387.00
250 G
$1,490.00

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$127.00
50 G
$387.00
250 G
$1,490.00

About This Item

Linear Formula:
CH3OCH=CHCOCH3
CAS Number:
Molecular Weight:
100.12
Beilstein:
2070991
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

Assay

90%

refractive index

n20/D 1.468 (lit.)

bp

200 °C (lit.)

density

0.982 g/mL at 25 °C (lit.)

functional group

ether
ketone

storage temp.

2-8°C

SMILES string

[H]\C(OC)=C(\[H])C(C)=O

InChI

1S/C5H8O2/c1-5(6)3-4-7-2/h3-4H,1-2H3/b4-3+

InChI key

VLLHEPHWWIDUSS-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-4-Methoxy-3-buten-2-one acts as substrate and undergoes zinc triflate-catalyzed Mukaiyama-Michael reaction with 3-TBSO-substituted vinyldiazoacetate to yield functionalized 3-keto-2-diazoalkanoates[1].

Application

trans-4-Methoxy-3-buten-2-one was used as starting reagent for enantioselective total synthesis of (-)-epibatidine[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Liu et al.
Organic letters, 12(19), 4304-4307 (2010-09-03)
Enedione-diazoesters formed from 3-TBSO-2-diazo-3-butenoates undergo base-catalyzed pericyclization that with dinitrogen extrusion and methyl migration provide a novel and efficient route to 2-carboalkoxyresorcinols. Intercepting the intermediate enolate anion with methyl vinyl ketone leads to the corresponding 4-substituted 2-carboalkoxyresorcinol and suggests generalization
Bifunctional thiourea-catalyzed enantioselective double Michael reaction of ?, d-unsaturated ?-ketoester to nitroalkene: asymmetric synthesis of (-)-epibatidine.
Hoashi Y, et al.
Tetrahedron Letters, 45(50), 9185-9188 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service