Skip to Content
Merck
All Photos(1)

Key Documents

172901

Sigma-Aldrich

3-(Trifluoromethyl)benzonitrile

99%

Synonym(s):

α,α,α-Trifluoro-m-tolunitrile, 3-Cyanobenzotrifluoride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$83.70

$83.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$83.70

About This Item

Linear Formula:
CF3C6H4CN
CAS Number:
Molecular Weight:
171.12
Beilstein:
1868102
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$83.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.4575 (lit.)

bp

189 °C (lit.)

mp

16-20 °C (lit.)

density

1.281 g/mL at 25 °C (lit.)

functional group

fluoro
nitrile

SMILES string

FC(F)(F)c1cccc(c1)C#N

InChI

1S/C8H4F3N/c9-8(10,11)7-3-1-2-6(4-7)5-12/h1-4H

InChI key

OGOBINRVCUWLGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(Trifluoromethyl)benzonitrile (α, α, α -trifluoro-m-tolunitrile) was used as a reference compound in the determination of the electron affinity of 1,3,5,7-cyclooctatetraene (COT)[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electron affinity of 1, 3, 5, 7-cyclooctatetraene determined by the kinetic method.
Denault JW, et al.
Journal of the American Society For Mass Spectrometry, 9(11), 1141-1145 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service