Skip to Content
Merck
All Photos(1)

Key Documents

165549

Sigma-Aldrich

2-Aminoanthraquinone

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9NO2
CAS Number:
Molecular Weight:
223.23
Beilstein:
745903
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

form

powder

mp

292-295 °C (dec.) (lit.)

solubility

ethanol: soluble(lit.)

SMILES string

Nc1ccc2C(=O)c3ccccc3C(=O)c2c1

InChI

1S/C14H9NO2/c15-8-5-6-11-12(7-8)14(17)10-4-2-1-3-9(10)13(11)16/h1-7H,15H2

InChI key

XOGPDSATLSAZEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Aminoanthraquinone was used in fabrication of high performance electrode of supercapacitor based on chemically modified graphene hydrogel. It was used in preparation of ordered assemblies of organic and biological molecules on gold(111) surfaces.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiong Wu et al.
Physical chemistry chemical physics : PCCP, 13(23), 11193-11198 (2011-05-13)
2-Aminoanthraquinone (AAQ) molecules were covalently grafted onto chemically modified graphene (CMG), and the AAQ functionalized CMG sheets were self-assembled into macroporous hydrogels for supercapacitor electrodes. The electrode based on the AAQ modified self-assembled graphene hydrogel (AQSGH) showed a high specific
Self-Organized Materials: From Organic molecules to Genetically Engineered Gold-Binding Proteins.
Zareie H, et al.
Nanoscience and Nanotechnology (2006)
2-Aminoanthraquinone.
Report on carcinogens : carcinogen profiles, 12, 36-37 (2011-08-11)
S V Gothoskar et al.
Medical biology, 58(5), 281-284 (1980-10-01)
The role of gonadal hormones in the nephrotoxicity of the hepatocarcinogen 2-aminoanthraquinone (2-AAQ) was investigated. Intact and castrated four week old Fischer rats of both sexes and castrated female rats with subcutaneous implants of testosterone propionate pellets were fed 2%
Proteinuria in the Fischer rat by feeding 2-aminoanthraquinone.
S V Gothoskar et al.
Medical biology, 58(6), 337-340 (1980-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service