Skip to Content
Merck
All Photos(2)

Key Documents

444650

Sigma-Aldrich

2-Bromomethyl-anthraquinone

97%

Synonym(s):

MAQ-Br

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$290.00

$290.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$290.00

About This Item

Empirical Formula (Hill Notation):
C15H9BrO2
CAS Number:
Molecular Weight:
301.13
Beilstein:
2276854
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$290.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

203 °C (dec.) (lit.)

functional group

bromo
ketone

SMILES string

BrCc1ccc2C(=O)c3ccccc3C(=O)c2c1

InChI

1S/C15H9BrO2/c16-8-9-5-6-12-13(7-9)15(18)11-4-2-1-3-10(11)14(12)17/h1-7H,8H2

InChI key

FCETXLHFBYOVCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some Organic-Inorganic Hybrid Compounds Based on iso-Thiuronium Cations and Lead Halide Anions.
Papavassiliou GC, et al.
Zeitschrift fur Naturforschung (2001)
Modified carbon nanoparticle-chitosan film electrodes: Physisorption versus chemisorption.
Rassaei L, et al.
Electrochimica Acta, 53(19), 5732-5738 (2008)
Graphene: Large scale chemical functionalization by cathodic means.
Jouikov V and Simonet J.
Electrochemical Communications, 46, 132-136 (2014)
Inclusion of anthraquinone derivatives by the cucurbit [7] uril host.
Sindelar V, et al.
New. J. Chem., 31(5), 725-728 (2007)
Antitermite properties of some anthraquinone derivatives.
Gupta BK and Sen-Sarma PK.
Holzforschung, 30(3), 57-58 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service