Skip to Content
Merck
All Photos(1)

Documents

16511

Sigma-Aldrich

3-Bromobutyric acid

≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHBrCH2COOH
CAS Number:
Molecular Weight:
167.00
Beilstein:
1720574
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

refractive index

n20/D 1.476

density

1.57 g/mL at 20 °C (lit.)

SMILES string

CC(Br)CC(O)=O

InChI

1S/C4H7BrO2/c1-3(5)2-4(6)7/h3H,2H2,1H3,(H,6,7)

InChI key

HAIUIAZIUDPZIE-UHFFFAOYSA-N

Application

3-Bromobutyric acid was used in the synthesis of β-butyrolactone, monomer for a naturally occurring polyester, D-poly-β-hydroxybutyrate. It was used to investigate the effect of halo acids on the activity of aspartate aminotransferase isoenzymes in their pyridoxamine form. It was also used in the synthesis of optically active β-lactones.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of poly-?-hydroxybutyrate. I. Synthesis of crystalline DL-poly-?-hydroxybutyrate from DL-?-butyrolactone.
Agostini DE, et al.
Journal of Polymer Science: Part A, General Papers, 9(10), 2775-2787 (1971)
Methods for the synthesis of optically active β-lactones (2-oxetanones).
Yang YW and Romo D.
Tetrahedron, 55(21), 6403-6434 (1999)
Selective inactivation of pyridoxamine form of aspartate aminotransferase by iodoacetate. Carboxymethylation of 4'-amino group of bound pyridoxamine 5'-phosphate.
Y Morino et al.
The Journal of biological chemistry, 253(17), 6026-6030 (1978-09-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service