Skip to Content
Merck
All Photos(1)

Key Documents

162841

Sigma-Aldrich

2-Methyl-1-naphthol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C10H6OH
CAS Number:
Molecular Weight:
158.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

64-66 °C (lit.)

storage temp.

−20°C

SMILES string

Cc1ccc2ccccc2c1O

InChI

1S/C11H10O/c1-8-6-7-9-4-2-3-5-10(9)11(8)12/h2-7,12H,1H3

InChI key

SRJCJJKWVSSELL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-1-naphthol undergoes oxidation with molecular oxygen to yield 2-methyl-1,4-naphthoquinone[1]. Mechanism of selective oxidation of 2-methyl-1-naphthol with H2O2 catalyzed by titanium single-site catalysts, TiO2-SiO2 aerogel and mesostructured hydrothermally stable titanium-silicate has been investigated by EPR spectroscopic technique[2].

Application

2-Methyl-1-naphthol was used in selective synthesis of vitamin K3 via liquid-phase oxidation using NbSBA-15 catalyst[3].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Selvaraj et al.
Dalton transactions (Cambridge, England : 2003), 41(32), 9633-9638 (2012-07-17)
Well hexagonally ordered NbSBA-15 catalysts synthesized by an efficient hydrothermal method were used, for the first time, for the selective synthesis of vitamin K(3) by liquid-phase oxidation of 2-methyl-1-naphthol (2MN1-OH) under various reaction conditions. The recyclable NbSBA-15 catalysts were also
EPR study on the mechanism of H2O2-based oxidation of alkylphenols over titanium single-site catalysts.
Zalomaeva OV, et al.
J. Mol. Catal. A: Chem., 277(1), 185-192 (2007)
Oxana A Kholdeeva et al.
The journal of physical chemistry. B, 115(42), 11971-11983 (2011-09-15)
Oxidation of 2-methyl-1-naphthol (MNL) with molecular oxygen proceeds efficiently under mild reaction conditions (3 atm O(2), 60-80 °C) in the absence of any catalyst or sensitizer and produces 2-methyl-1,4-naphthoquinone (MNQ, menadione, or vitamin K(3)) with selectivity up to 80% in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service