Skip to Content
Merck
All Photos(2)

Key Documents

162140

Sigma-Aldrich

Diethoxymethyl acetate

96%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$897.00

$897.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$897.00

About This Item

Linear Formula:
CH3COOCH(OCH2CH3)2
CAS Number:
Molecular Weight:
162.18
Beilstein:
1209886
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$897.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

96%

form

liquid

refractive index

n20/D 1.399 (lit.)

density

0.993 g/mL at 25 °C (lit.)

functional group

ester
ether

storage temp.

2-8°C

SMILES string

CCOC(OCC)OC(C)=O

InChI

1S/C7H14O4/c1-4-9-7(10-5-2)11-6(3)8/h7H,4-5H2,1-3H3

InChI key

IRUNKQSGDBYUDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethoxymethyl acetate reacts with 5-amino-6-ribitylaminouracil hydrochloride to yield 6-dioxo-(1H,3H)-9-N-ribitylpurine[1].

Application

Diethoxymethyl acetate was used in the synthesis of D-mannosyl, D-galactosyl and D-glucosyl theophylline nucleosides[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Cushman et al.
Bioorganic & medicinal chemistry, 6(4), 409-415 (1998-05-23)
2,6-Dioxo-(1H,3H)-9-N-ribitylpurine (6) and 2,6-dioxo-(1H,3H)-8-aza-9-N-ribitylpurine (7) have been synthesized and evaluated as inhibitors of lumazine synthase and riboflavin synthase. Reaction of 5-amino-6-ribitylaminouracil hydrochloride (8) with diethoxymethyl acetate (9) afforded the purine 6, while diazotization of 8 afforded the 8-aza purine 7.
Rodrigo Rico-Gómez et al.
Carbohydrate research, 343(5), 855-864 (2008-02-16)
The synthesis of D-mannosyl, D-galactosyl and D-glucosyl theophylline nucleosides by diethoxymethyl acetate (DEMA)-induced cyclization of 4-amino-5-glycosylideneimino-1,3-dimethyluracil is reported. 8-Methyltheophylline derivatives of the same sugars were also prepared by Ac(2)O/H(+)-induced cyclization of their imine precursors. This approach has allowed beta-D-mannopyranosyl-, alpha-D-galactofuranosyl-

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service