Skip to Content
Merck
All Photos(1)

Key Documents

8.43948

Sigma-Aldrich

1,1,2,2-Tetrachloroethane

for synthesis

Synonym(s):

1,1,2,2-Tetrachloroethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2Cl4
CAS Number:
Molecular Weight:
167.85
MDL number:
UNSPSC Code:
12352101
EC Index Number:
201-197-8
NACRES:
NA.22
grade:
for synthesis
synthesis grade
vapor pressure:
6.4 hPa ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

6.4 hPa ( 20 °C)

Assay

≥99.0% (GC)

form

liquid

potency

200 mg/kg LD50, oral (Rat)
3990 mg/kg LD50, skin (Rabbit)

dilution

(for synthesis)

mp

-42.5 °C

solubility

2830 g/L

density

1.59 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

ClC(Cl)C(Cl)Cl

InChI

1S/C2H2Cl4/c3-1(4)2(5)6/h1-2H

InChI key

QPFMBZIOSGYJDE-UHFFFAOYSA-N

Application


  • Activity coefficients and excess Gibbs energy functions of acetophenone with 1, 2-dichloroethane and 1, 1, 2, 2-tetrachloroethane binary mixtures: Provides data on the thermodynamic properties of 1,1,2,2-Tetrachloroethane in chemical mixtures, useful for chemical engineering applications (Anila et al., 2016).


Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.593 - 1.598
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service