Skip to Content
Merck
All Photos(1)

Key Documents

160792

Sigma-Aldrich

11-Phenoxyundecanoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5O(CH2)10CO2H
CAS Number:
Molecular Weight:
278.39
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

bp

217-222 °C/3 mmHg (lit.)

mp

77-78 °C (lit.)

solubility

chloroform: soluble 25 mg/mL, clear (Colorless to orange)

SMILES string

OC(=O)CCCCCCCCCCOc1ccccc1

InChI

1S/C17H26O3/c18-17(19)14-10-5-3-1-2-4-6-11-15-20-16-12-8-7-9-13-16/h7-9,12-13H,1-6,10-11,14-15H2,(H,18,19)

InChI key

FRSQLPPSRJNREN-UHFFFAOYSA-N

Application

11-Phenoxyundecanoic acid was employed as carbon source in the biosynthesis of aromatic copolyesters by isolated Pseudomonas putida BM01.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Song et al.
Applied and environmental microbiology, 62(2), 536-544 (1996-02-01)
Two types of novel aromatic copolyesters were synthesized from 11-phenoxyundecanoic acid (11-POU) as the sole carbon source and the cosubstrates 11-POU and octanoate, respectively, by isolated Pseudomonas putida BM01 that is known to accumulate high concentrations of medium-chain-length polyesters. Insoluble

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service