Skip to Content
Merck
All Photos(1)

Key Documents

158984

Sigma-Aldrich

1,3-Diacetylbenzene

97%

Synonym(s):

1,1′-(1,3-Phenylene)bis[ethanone], 1,3-Bis(1-oxoethyl)benzene, 1-(3-Acetylphenyl)ethanone, 2,6-Diacetylbenzene, 3-Acetylacetophenone, m-Acetylacetophenone, m-Diacetylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$265.00

$265.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$265.00

About This Item

Linear Formula:
C6H4(COCH3)2
CAS Number:
Molecular Weight:
162.19
Beilstein:
2042511
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$265.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

97%

form

crystals

bp

150-155 °C/15 mmHg (lit.)

mp

28-32 °C (lit.)

functional group

ketone

storage temp.

2-8°C

SMILES string

CC(=O)c1cccc(c1)C(C)=O

InChI

1S/C10H10O2/c1-7(11)9-4-3-5-10(6-9)8(2)12/h3-6H,1-2H3

InChI key

VCHOFVSNWYPAEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Diacetylbenzene is an δ-diketone. It induces neuropathological changes in the rodent central and peripheral nervous systems.[1]

Application

1,3-Diacetylbenzene was used in the preparation of polyhydroxylated analogs.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Artico et al.
Journal of medicinal chemistry, 41(21), 3948-3960 (1998-10-10)
Various cinnammoyl-based structures were synthesized and tested in enzyme assays as inhibitors of the HIV-1 integrase (IN). The majority of compounds were designed as geometrically or conformationally constrained analogues of caffeic acid phenethyl ester (CAPE) and were characterized by a
Min-Sun Kim et al.
Toxicology and applied pharmacology, 183(1), 55-65 (2002-09-10)
The gamma-diketone analogs 1,2-diacetylbenzene (1,2-DAB) and 2,5-hexanedione (2,5-HD), but not the delta-diketone 1,3-diacetylbenzene (1,3-DAB) or the beta-diketone 2,4-hexanedione, induce neuropathological changes in the rodent central and peripheral nervous systems. The molecular targets of these neurotoxic aromatic and aliphatic gamma-diketones, and
Tim Goelen et al.
Pest management science, 77(2), 930-938 (2020-09-26)
Recent studies have shown that microorganisms emit volatile compounds that affect insect behaviour. However, it remains largely unclear whether microbes can be exploited as a source of attractants to improve biological control of insect pests. In this study, we used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service