Skip to Content
Merck
All Photos(2)

Key Documents

261432

Sigma-Aldrich

Methyl coumalate

98%

Synonym(s):

Methyl 2-oxo-2H-pyran-5-carboxylate, Methyl 2-pyrone-5-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$468.00

$468.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$468.00

About This Item

Empirical Formula (Hill Notation):
C7H6O4
CAS Number:
Molecular Weight:
154.12
Beilstein:
126301
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$468.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

bp

178-180 °C/60 mmHg (lit.)

mp

65-67 °C (lit.)

functional group

ester

SMILES string

COC(=O)C1=COC(=O)C=C1

InChI

1S/C7H6O4/c1-10-7(9)5-2-3-6(8)11-4-5/h2-4H,1H3

InChI key

HHWWWZQYHPFCBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl coumalate is a 2-pyrone and acts as dienophile in Diels-Alder reaction. It reacts with 1,3-butadienes at 100°C to yield tetrahydrocoumarins and 4-methoxycarbonyltricyclo[3.2.1.02,7]octenes. It also reacts with cyclohexadiene to afford tetrahydronaphthalene-2-carboxylate.[1] It undergoes Diels-Alder reaction with unactivated alkenes to afford para-substituted adducts.[2]

Application

Methyl coumalate has been used as reagent in phosphine-catalyzed [4+3] annulation of modified allylic carbonates.[3] It was also used in the preparation of 7-carboxyquinolizinium derivatives.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aromatics from pyrones: para-substituted alkyl benzoates from alkenes, coumalic acid and methyl coumalate.
Kraus GA, et al.
Green Chemistry, 13(10), 2734-2736 (2011)
Diels-Alder reaction of methyl coumalate with 1, 3-dienes.
Imagawa T, et al.
Tetrahedron, 30(14), 2227-2231 (1974)
Suqing Zheng et al.
Organic letters, 11(17), 3978-3981 (2009-08-13)
A phosphine-catalyzed [4 + 3] annulation of modified allylic carbonates with methyl coumalate was developed. This strategy offered a powerful method for the construction of bicyclo[3.2.2]nonadiene skeleton with high stereoselectivity.
Quinolizinium Compounds by Cyclization of Pyridones from Methyl Coumalate and ?-Phenylethylamines.
Wiley RH, et al.
Journal of the American Chemical Society, 75(18), 4482-4484 (1953)

Articles

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service