Skip to Content
Merck
All Photos(2)

Key Documents

158658

Sigma-Aldrich

Isobutyl formate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
102.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

vapor density

3.52 (vs air)

Assay

97%

form

liquid

autoignition temp.

608 °F

expl. lim.

8.9 %

refractive index

n20/D 1.385 (lit.)

bp

98.4 °C (lit.)

density

0.885 g/mL at 25 °C (lit.)

SMILES string

CC(C)COC=O

InChI

1S/C5H10O2/c1-5(2)3-7-4-6/h4-5H,3H2,1-2H3

InChI key

AVMSWPWPYJVYKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isobutyl formate was used in the peptide formation during the synthesis of pteroyl-(γ-L-glutamyl)n-L-glutamic acid (upto n=6).[1]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The solid-phase synthesis of polyglutamates of folic acid.
C L Krumdieck et al.
Biochemistry, 8(4), 1568-1572 (1969-04-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service