Skip to Content
Merck
All Photos(1)

Key Documents

157643

Sigma-Aldrich

3-Methylcyclopentanone

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$154.00

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$154.00

About This Item

Linear Formula:
CH3C5H7(=O)
CAS Number:
Molecular Weight:
98.14
Beilstein:
1305074
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

145 °C (lit.)

density

0.913 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC1CCC(=O)C1

InChI

1S/C6H10O/c1-5-2-3-6(7)4-5/h5H,2-4H2,1H3

InChI key

AOKRXIIIYJGNNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methylcyclopentanone is a monocyclic ketone and its optical rotatory dispersion has been studied under isolated and solvated conditions to explore the role of ring size/morphology.[1] The vibrationally resolved electronic circular dichroism (ECD) spectra of (R)-(+)-3-methylcyclopentanone in gas phase was evaluated by density functional theory.[2]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Lin et al.
The journal of physical chemistry. A, 112(48), 12401-12411 (2008-11-13)
The vibrationally resolved electronic circular dichroism (ECD) spectra of the two dominant conformers of (R)-(+)-3-methylcyclopentanone in gas phase are computed by density functional response theory, with a full account of Franck-Condon and Herzberg-Teller vibrational contributions at the harmonic level. Proper
Kajetan Koperwas et al.
Scientific reports, 6, 36934-36934 (2016-11-25)
When we cool down a liquid below the melting temperature, it can either crystallize or become supercooled, and then form a disordered solid called glass. Understanding what makes a liquid to crystallize readily in one case and form a stable
Priyanka Lahiri et al.
The journal of physical chemistry. A, 117(47), 12382-12400 (2013-11-15)
The optical rotatory dispersion of two monocyclic ketones, (R)-3-methylcyclopentanone [R-3MCP] and (R)-3-methylcyclohexanone [R-3MCH], has been investigated under isolated and solvated conditions to explore the role of ring size/morphology and to elucidate the impact of environmental perturbations. Vapor-phase measurements of specific

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service