Skip to Content
Merck
All Photos(1)

Key Documents

154466

Sigma-Aldrich

3,4-Dimethoxystyrene

technical grade, contains 1% hydroquinone as inhibitor

Synonym(s):

1,2-Dimethoxy-4-vinylbenzene, 3,4-Dimethoxystyrol, 4-Ethenyl-1,2-dimethoxybenzene, 4-Vinylveratrole

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$227.00
10 G
$388.00

$227.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$227.00
10 G
$388.00

About This Item

Linear Formula:
(CH3O)2C6H3CH=CH2
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$227.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

grade

technical grade

form

liquid

contains

1% hydroquinone as inhibitor

refractive index

n20/D 1.571 (lit.)

bp

120-125 °C/10 mmHg (lit.)

density

1.109 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc(C=C)cc1OC

InChI

1S/C10H12O2/c1-4-8-5-6-9(11-2)10(7-8)12-3/h4-7H,1H2,2-3H3

InChI key

NJXYTXADXSRFTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Characteristic Scent from the Tahitian Liverwort, Cyathodium foetidissimum.: The study discusses the unique scent compounds identified in the Tahitian liverwort, with a mention of 3,4-Dimethoxystyrene as a component. The research highlights the significance of such compounds in the field of biochemistry and their potential applications in various biochemical processes (Sakurai et al., 2018).

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simrone K Gill et al.
Acta biomaterialia, 63, 326-335 (2017-09-12)
Nature provides many interesting examples of adhesive strategies. Of particular note, the protein glue secreted by marine mussels delivers high adhesion in wet and dynamic environments owing to existence of catechol moieties. As such, this study focuses on denture fixatives

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service