Skip to Content
Merck
All Photos(1)

Key Documents

151165

Sigma-Aldrich

Thallium(III) acetate

Synonym(s):

Tl(OAc)3, thallic acetate, tris(acetate)thallium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)3Tl
CAS Number:
Molecular Weight:
381.52
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: thallium
reagent type: catalyst

concentration

51.0-56.0% (TL, ICP)

impurities

≤10% acetic acid (may contain)

mp

182 °C (dec.) (lit.)
182 °C (dec.) (lit.)

SMILES string

[TlH].CC(O)=O

InChI

1S/3C2H4O2.Tl/c3*1-2(3)4;/h3*1H3,(H,3,4);/q;;;+3/p-3

InChI key

SMRRYUGQTFYZGD-UHFFFAOYSA-K

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Eybl et al.
Ceskoslovenska farmacie, 41(7-8), 243-245 (1992-10-01)
Experiments carried out on male mice (ICR) demonstrated a protective effect of premedication with the preparation Sho-Saiko-To (TJ 9, Tsumura and Comp.) against the hepatotoxic effects of CCl4 and T1-acetate, which were manifested by increased peroxidation of lipids and increased
R Barroso-Moguel et al.
Archives of medical research, 23(3), 129-133 (1992-01-01)
An i.p. injection of a solution of thallium acetate in deionized water at a dose of 32 mg/kg, in 24-h-old rats, produces morphological and biochemical alterations in both cartilaginous and osseous tissues. From the beginning, there are alterations in the
M Csicsaky et al.
Toxicology, 49(1), 121-129 (1988-04-01)
The toxic effect of thallium added to the bath solution was studied with intra- and extracellular recordings from mammalian nerve-muscle preparations. To elucidate the target region, 3 different functional parameters were studied: (1) Post-synaptic endplate potentials (EPPs) resulting from evoked
R Barroso-Moguel et al.
Journal of applied toxicology : JAT, 16(5), 385-389 (1996-09-01)
A histopathological study, with a light microscope, of experimental neuromyopathy produced by thallotoxicosis was undertaken in 40 newborn Wistar rats. Treatment consisted of a single i.p. injection of an aqueous solution of 16 mg kg-1 thallium(I) acetate 1 day after
J G Tully et al.
Sexually transmitted diseases, 10(4 Suppl), 256-260 (1983-10-01)
Specimens from the cervix or posterior vaginal fornix of 50 women attending a venereal disease clinic and from the urethra of 50 men with nongonococcal urethritis were cultured for Mycoplasma hominis. Recovery of the organism in two conventional mycoplasma culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service