Skip to Content
Merck
All Photos(4)

Documents

T8266

Sigma-Aldrich

Thallium(I) acetate

≥99%

Synonym(s):

Acetic acid thallium(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOTl
CAS Number:
Molecular Weight:
263.43
Beilstein:
3693914
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99%

form

powder

reaction suitability

core: thallium
reagent type: catalyst

SMILES string

CC(=O)O[Tl]

InChI

1S/C2H4O2.Tl/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

HQOJMTATBXYHNR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Thallium (I) acetate can be prepared by dissolving thallium (III) oxide in acetic acid and acetic anhydride and stirring at 80-90oC. After filtration, the solution cooled and the product is recrystallized. A study reports that a large number of mycoplasmas and acholeplasmas can grow in its presence at concentration of 1 mg/ml.

Application

Commonly used in the laboratories as a selective element against gram-negative bacteria in selective media for the detection of mollicutes. It may be used in synthesizing thallium derivatives.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Properties and Structures of Thallium (I) Derivatives of Aromatic Diols.
El-Hadad AA, et al.
Inorganic Chemistry, 34(1), 120-123 (1995)
Selective media and faecal streptococci recovery: a review.
F A Saleh
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. Zweite naturwissenschaftliche Abteilung: Mikrobiologie der Landwirtschaft der Technologie und des Umweltschutzes, 135(2), 130-144 (1980-01-01)
T Ohyashiki et al.
Biochimica et biophysica acta, 858(2), 294-300 (1986-06-26)
The effect of alpha-tocopherol on the lipid fluidity of porcine intestinal brush-border membranes was studied using pyrene as a fluorescent probe. Addition of alpha-tocopherol to the medium decreased fluorescence intensity and lifetime, but increased the fluorescence polarization of pyrene-labeled membranes.
D C Dragon et al.
Letters in applied microbiology, 33(2), 100-105 (2001-07-27)
To investigate methods of improving anthrax spore detection with PLET. Comparisons were made of PLET and blood-supplemented PLET to recover and distinguish spores of a variety of Bacillus species. Heat and ethanol purification of spores, and spore extraction from soil
Chee-Fah Chia et al.
Annals of the New York Academy of Sciences, 1042, 523-530 (2005-06-21)
Thallium acetate is a known neurotoxic agent. In this study, we investigated the mechanisms by which thallium acetate induces cell cycle arrest and cell apoptosis in cultured LC6 glioma cells. Exposure of C6 glioma cells to thallium acetate decreased cell

Articles

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service