Skip to Content
Merck
All Photos(1)

Key Documents

142158

Sigma-Aldrich

1,3-Diphenylurea

98%

Synonym(s):

N,N′-Diphenylurea, Carbanilide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$64.80
100 G
$195.00

$64.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$64.80
100 G
$195.00

About This Item

Linear Formula:
(C6H5NH)2CO
CAS Number:
Molecular Weight:
212.25
Beilstein:
782650
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$64.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder

bp

262 °C (lit.)

mp

239-241 °C (lit.)

solubility

pyridine: soluble 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

functional group

amine

SMILES string

O=C(Nc1ccccc1)Nc2ccccc2

InChI

1S/C13H12N2O/c16-13(14-11-7-3-1-4-8-11)15-12-9-5-2-6-10-12/h1-10H,(H2,14,15,16)

InChI key

GWEHVDNNLFDJLR-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1,3-Diphenylurea is a cytokinin compound present in fruit and vegetables[1].

Biochem/physiol Actions

1,3-Diphenylurea is metabolized by a moderate halophilic Marinobacter sp. isolated from a contaminated ephemeral desert stream bed in Negev desert[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian R Sørensen et al.
FEMS microbiology letters, 213(2), 199-204 (2002-08-09)
A moderate halophilic Marinobacter sp. (designated strain DPUZ) able to metabolize 1,3-diphenylurea (DPU) was isolated from a contaminated ephemeral desert stream bed near an industrial complex in the northern part of the Negev Desert (Israel). Metabolism of DPU was accompanied
N Campillo et al.
Talanta, 116, 376-381 (2013-10-24)
The paper presents a novel approach for the determination of three cytokinin compounds, thidiazuron (TDZ), 1,3-diphenylurea (1,3-DPU) and forchlorfenuron (CPPU), in fruit and vegetables samples using liquid chromatography with electrospray ionization and time-of-flight mass spectrometry (LC-ESI-TOFMS). Analytes were extracted from
Joshua R Brown et al.
Bioorganic & medicinal chemistry, 19(18), 5585-5595 (2011-08-16)
The treatment of tuberculosis is becoming more difficult due to the ever increasing prevalence of drug resistance. Thus, it is imperative that novel anti-tuberculosis agents, with unique mechanisms of action, be discovered and developed. The direct anti-tubercular testing of a
Nadezhda German et al.
European journal of medicinal chemistry, 43(11), 2453-2463 (2008-03-25)
Bacterial efflux pump systems contribute to antimicrobial resistance in pathogenic bacteria. The co-administration of bacterial efflux pump inhibitors with antibiotics is being pursued to overcome efflux-mediated resistance to antibiotics. In this study we investigated a strategy for converting broad-spectrum efflux
Paul Bamborough et al.
Journal of medicinal chemistry, 54(14), 5131-5143 (2011-06-28)
A kinase-focused screening set of fragments has been assembled and has proved successful for the discovery of ligand-efficient hits against many targets. Here we present some of our general conclusions from this exercise. Notably, we present the first profiling results

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service