Merck
Selectivity of kinase inhibitor fragments.
Journal of medicinal chemistry (2011-06-28)
Paul Bamborough, Murray J Brown, John A Christopher, Chun-wa Chung, Geoff W Mellor
ABSTRACT
A kinase-focused screening set of fragments has been assembled and has proved successful for the discovery of ligand-efficient hits against many targets. Here we present some of our general conclusions from this exercise. Notably, we present the first profiling results for literature fragments that have previously been used as starting points for optimization against individual kinases. We consider the importance of screening format and the extent to which selectivity is helpful in selecting fragments for progression. Results are also outlined for fragments targeting the DFG-out conformation and for atypical kinases such as PIM1 and lipid kinases.
Sigma-Aldrich
7-Azaindole, 98%
Sigma-Aldrich
1,3-Diphenylurea, 98%
Sigma-Aldrich
Diphenylamine, ACS reagent, ≥99%
Supelco
Diphenylamine, PESTANAL®, analytical standard
Sigma-Aldrich
Diphenylamine, ReagentPlus®, 99%
Sigma-Aldrich
Adenine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Diphenylamine, puriss. p.a., suitable for redox indicator, ACS reagent, reag. Ph. Eur., ≥98% (GC)
Sigma-Aldrich
Adenine, BioReagent, suitable for plant cell culture, ≥99%