Skip to Content
Merck
All Photos(1)

Key Documents

139149

Sigma-Aldrich

Trimethylphenylammonium iodide

99%

Synonym(s):

Phenyltrimethylammonium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(I)C6H5
CAS Number:
Molecular Weight:
263.12
Beilstein:
3916752
EC Number:
MDL number:
UNSPSC Code:
12352000

Assay

99%

mp

227 °C (subl.) (lit.)

SMILES string

[I-].C[N+](C)(C)c1ccccc1

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neera Singh
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 41(1), 17-29 (2006-01-06)
The study aims to prepare the organoclay complexes of metolachlor and metribuzin so as to reduce their downward mobility in soil profile. The organoclays were preadsorbed with phenyltrimethylammonium (PTMA) (50% of cation exchange capacity [CEC]) and hexadecyltrimethylammonium (HDTMA) (100% of
Godfrey Lisk et al.
Molecular pharmacology, 71(5), 1241-1250 (2007-02-09)
Human red blood cells infected with the malaria parasite Plasmodium falciparum have markedly increased permeabilities to diverse organic and inorganic solutes. The plasmodial surface anion channel (PSAC), recently identified with electrophysiological methods, contributes to the uptake of many small solutes.
Alec N Salt et al.
Hearing research, 283(1-2), 14-23 (2011-12-20)
It has been widely believed that drug entry from the middle ear into perilymph occurs primarily via the round window (RW) membrane. Entry into scala vestibuli (SV) was thought to be dominated by local, inter-scala communication between scala tympani (ST)
Petr Gebauer et al.
Electrophoresis, 29(5), 1067-1076 (2008-01-26)
In electrophoresis, the introduction of a sample into the BGE creates two new zone boundaries, the front and rear boundary of the sample zone which is sandwiched between the BGE. During an electrophoresis run, these boundaries move and split into
Alec N Salt et al.
Hearing research, 182(1-2), 24-33 (2003-09-02)
Our understanding of the perilymph kinetics of drugs depends largely on data obtained by the analysis of perilymph samples. Although a number of studies have demonstrated qualitatively that perilymph samples may be contaminated by cerebrospinal fluid (CSF), and some investigations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service