Skip to Content
Merck
All Photos(1)

Key Documents

13815

Sigma-Aldrich

1-Benzylpiperazine

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2
CAS Number:
Molecular Weight:
176.26
Beilstein:
141624
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (GC)

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada

refractive index

n20/D 1.547 (lit.)
n20/D 1.547

mp

17-20 °C

density

1.014 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

C1CN(CCN1)Cc2ccccc2

InChI

1S/C11H16N2/c1-2-4-11(5-3-1)10-13-8-6-12-7-9-13/h1-5,12H,6-10H2

InChI key

IQXXEPZFOOTTBA-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

1-Benzylpiperazine has stimulant and euphoric properties[1]. It is commonly used constituent of party pill drugs[2]. It is a potential antidepressant and is an active metabolite of the withdrawn antidepressant drug piberaline[3].

Application

1-Benzylpiperazine was used to study the inhibition of various acetylcholinesterases by 7-ethyl-10-[4-(1-piperidino)-1-piperidino]carbonyloxycamptothecin and related compounds[4].

Other Notes

Sales restrictions may apply

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

253.4 °F - closed cup

Flash Point(C)

123 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Elliott et al.
Journal of analytical toxicology, 32(2), 172-177 (2008-03-13)
Piperazines such as 1-benzylpiperazine (BZP), 1-(3-trifluromethylphenyl)piperazine (3-TFMPP), and 1-(3-chlorophenyl)piperazine (3-CPP) have become recent drugs of abuse. With stimulant effects comparable to amphetamines but with a lower potency and differential global scheduling status, they have been sold as a supposed legal
Márcia Sá Monteiro et al.
Archives of toxicology, 87(6), 929-947 (2013-05-21)
N-benzylpiperazine (BZP) has become popular among recreational drug users as the major active ingredient of "party pills" due to its stimulant and euphoric properties. Before BZP legal restrictions, these pills were sold as a safe and legal alternative to classical
The crystal structure of the complex of the anticancer prodrug 7-ethyl-10-[4-(1-piperidino)-1-piperidino]-carbonyloxycamptothecin (CPT-11) with Torpedo californica acetylcholinesterase provides a molecular explanation for its cholinergic action.
Harel M, et al.
Molecular Pharmaceutics, 67(6), 1874-1881 (2005)
Ellen I Dixon et al.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience, 73, 32-40 (2018-12-28)
From 30 days after birth until the completion of the study, male and female rats were caged in same-sexed twos or threes either with (enriched cages, EC) or without several objects for them to explore (standard cages, SC). From 41
David M Wood et al.
Therapeutic drug monitoring, 34(4), 363-367 (2012-06-08)
Since the turn of the 21st century, there has been an increase in the availability and use of novel psychoactive substances (also known as "legal highs") across Europe. Currently, there is limited information available on the potential acute toxicity (harms)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service