Skip to Content
Merck
All Photos(1)

Key Documents

A45564

Sigma-Aldrich

2-Amino-5-chlorobenzophenone

98%

Synonym(s):

4-Chloro-2-benzoylaniline

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$59.50
100 G
$82.90

$59.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$59.50
100 G
$82.90

About This Item

Linear Formula:
H2NC6H3(Cl)COC6H5
CAS Number:
Molecular Weight:
231.68
Beilstein:
475640
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$59.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

mp

96-98 °C (lit.)

SMILES string

Nc1ccc(Cl)cc1C(=O)c2ccccc2

InChI

1S/C13H10ClNO/c14-10-6-7-12(15)11(8-10)13(16)9-4-2-1-3-5-9/h1-8H,15H2

InChI key

ZUWXHHBROGLWNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

388.4 °F - closed cup

Flash Point(C)

198 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Swetlana Heinrich et al.
European journal of medicinal chemistry, 46(4), 1331-1342 (2011-02-25)
Previously we described a series of 5-acylaminobenzophenones with considerable antimalarial activity. Unfortunately, most compounds also displayed high cytotoxicity resulting in low selectivity towards malaria parasites. Through the replacement of the 5-acylamino moiety by simple chlorine and further modifications of the
F J Rodríguez et al.
Journal of chromatography, 578(1), 146-151 (1992-07-01)
A high-performance liquid chromatographic method with electrochemical detection has been developed for the determination of three aminohalogenbenzophenones: 2-amino-2',5-dichlorobenzophenone, 2-amino-5-chlorobenzophenone and 2-amino-5-bromo-2'-fluorobenzophenone, metabolites of benzodiazepinooxazoles and other psychotropic drugs. A mobile phase of methanol-water (65:35), containing 5 mM KH2PO4 appeared to
R Jain
Journal of chromatography, 615(2), 365-368 (1993-06-02)
A direct densitometric method for determination of diazepam and its metabolites in urine was developed. The proposed procedure involves acid hydrolysis of urine specimens, thereby converting diazepam and its metabolites into benzophenones [2-methylamino-5-chlorobenzophenone (MACB) and 2-amino-5-chlorobenzophenone (ACB)]. It is followed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service