Skip to Content
Merck
All Photos(1)

Key Documents

135100

Sigma-Aldrich

Triphenylphosphine sulfide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3PS
CAS Number:
Molecular Weight:
294.35
Beilstein:
645872
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

161-163 °C (lit.)

solubility

isooctane: soluble

SMILES string

S=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H15PS/c20-19(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

VYNGFCUGSYEOOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triphenylphosphine sulfide (TPPS) forms TPPS·3I2 adduct with I2 molecules.

Application

Triphenylphosphine sulfide was used as internal standard in derivatization method for determination of trace levels of elemental sulphur in gasoline using flame ionization detection. It was also used for extraction of Pd(II) from hydrochloric acid solutions containing small amounts of SCN-.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Pauls
Journal of chromatographic science, 48(4), 283-288 (2010-04-24)
Several years ago, the presence of elemental sulfur in gasoline became a significant issue for the automotive and fuel industries. In several incidents, elemental sulfur at trace levels led to the corrosion of silver alloy fuel sensing elements in automobile
SCN- effect on the palladium (II) transfer in two and three phases systems using triphenylphosphine sulfide as a carrier.
Antico E, et al.
Reactive functional Polymers, 28(2), 103-109 (1996)
A new assembly of diiodine molecules at the triphenylphosphine sulfide template.
Devillanova FA.
J. Chem. Soc., Dalton Trans., 17, 3069-3073 (1999)
Shuli Yin et al.
Nanoscale, 12(27), 14863-14869 (2020-07-08)
The development of doped noble metal catalysts with nonmetal elements to improve the catalytic performance toward the oxygen reduction reaction (ORR) is significant for proton exchange membrane fuel cell technology. Here, we report a one-pot for dual-nonmetal-doping strategy for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service