Skip to Content
Merck
All Photos(1)

Key Documents

135046

Sigma-Aldrich

4-Nitrophenyl trimethylacetate

98%

Synonym(s):

4-Nitrophenyl pivalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCO2C6H4NO2
CAS Number:
Molecular Weight:
223.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

92-95 °C (lit.)

functional group

ester
nitro

SMILES string

CC(C)(C)C(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C11H13NO4/c1-11(2,3)10(13)16-9-6-4-8(5-7-9)12(14)15/h4-7H,1-3H3

InChI key

QADVJDGFQGNSIF-UHFFFAOYSA-N

General description

4-Nitrophenyl trimethylacetate undergoes hydrolysis catalysed by cytoplasmic aldehyde dehydrogenase in presence of NAD+, NADH, propionaldehyde, chloral hydrate, diethylstilboestrol and p-nitrobenzaldehyde (modifiers).

Application

4-Nitrophenyl trimethylacetate was used as sensitive probe to study the catalytic activity of carboxypeptidase Y.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The trimethylacetyl-transglutaminase complex.
J E Folk
Methods in enzymology, 87, 36-42 (1982-01-01)
H C Margolis et al.
The Journal of biological chemistry, 253(21), 7891-7897 (1978-11-10)
In the course of our further investigation of the active site titration of carboxypeptidase Y, using 4-nitrophenyl trimethylacetate, we have found that carboxypeptidase Y can be isolated in different molecular forms. Carboxypeptidase Y obtained from Fleischmann baker's yeast has a
T M Kitson
The Biochemical journal, 257(2), 573-578 (1989-01-15)
The effects of modifiers (NAD+, NADH, propionaldehyde, chloral hydrate, diethylstilboestrol and p-nitrobenzaldehyde) on the hydrolysis of p-nitrophenyl (PNP) pivalate (PNP trimethylacetate) catalysed by cytoplasmic aldehyde dehydrogenase are reported. In each case a different inhibition pattern is obtained to that observed
Studies of the esterase activity of cytosolic aldehyde dehydrogenase using sterically hindered and cyclic substrates.
K E Kitson et al.
Advances in experimental medicine and biology, 372, 45-52 (1995-01-01)
Roall Espersen et al.
Applied microbiology and biotechnology, 104(6), 2513-2522 (2020-01-29)
Two proteases, named C- and T-like proteases, respectively, were purified from the culture supernatant of Amycolatopsis keratinophila subsp. keratinophila D2T grown on a keratinous slaughterhouse by-product of pig bristles and nails as sole nitrogen and carbon source. The two proteases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service