Skip to Content
Merck
All Photos(1)

Key Documents

B3253

Sigma-Aldrich

Butyrylthiocholine iodide

≥98%

Synonym(s):

(2-Mercaptoethyl)trimethylammonium iodide butyrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$86.90
25 G
$191.00

$86.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$86.90
25 G
$191.00

About This Item

Linear Formula:
(CH3)3N(I)CH2CH2SCOCH2CH2CH3
CAS Number:
Molecular Weight:
317.23
Beilstein:
3729509
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$86.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98%

form

powder

mp

171-174 °C (lit.)

storage temp.

−20°C

SMILES string

[I-].CCCC(=O)SCC[N+](C)(C)C

InChI

1S/C9H20NOS.HI/c1-5-6-9(11)12-8-7-10(2,3)4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

WEQAAFZDJROSBF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Butyrylthiocholine iodide is a sulfur-containing analog of butyrylcholine. It is used as a reagent for the determination of butyrylcholinesterase activity.

Application

  • Label-Free and Ultrasensitive Detection of Butyrylcholinesterase: A study demonstrated the use of Mn(II)-based electron spin resonance spectroscopy for the ultrasensitive detection of butyrylcholinesterase, using Butyrylthiocholine iodide as a substrate to quantify enzyme activity in the presence of organophosphorus pesticides, crucial for biochemical assay applications (Tang et al., 2022).
  • Novel Nanozyme for Biosensing: Research developed a Co, N co-doped porous carbon-based nanozyme, demonstrating its utility as an oxidase mimic for fluorescence and colorimetric biosensing of butyrylcholinesterase, employing Butyrylthiocholine iodide as a key substrate, relevant in enzyme kinetics analysis (Sun et al., 2022).
  • Detection System for Anti-Alzheimer′s Drug Screening: A fluorescent platform was constructed using copper nanoclusters and MnO2 nanosheets for the detection of butyrylcholinesterase activity, utilizing Butyrylthiocholine iodide, which may facilitate the screening of anti-Alzheimer′s drugs and probe cholinergic system interactions (Chen et al., 2022).
  • Dual-Channel Detection of Butyrylcholinesterase: A study introduced bifunctional metal-organic frameworks with integrated fluorescence and oxidase activities, developed for dual-channel detection of butyrylcholinesterase using Butyrylthiocholine iodide, enhancing methodologies in biochemical assays (Wang et al., 2022).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ganiyu Oboh et al.
Neurotoxicology, 77, 12-19 (2019-12-15)
Many plant foods are rich sources of rutin, a flavonoid with many biological activities and health benefits. Exposure to cadmium has been implicated in neurotoxicity and cognitive dysfunction in animal models. However, there is a dearth of information on the
Oya Unsal-Tan et al.
MedChemComm, 10(6), 1018-1026 (2019-07-16)
A novel series of 2-pyrazoline derivatives were designed, synthesized, and evaluated for cholinesterase (ChE) inhibitory, Aβ anti-aggregating and neuroprotective activities. Among these, 3d, 3e, 3g, and 3h were established as the most potent and selective BChE inhibitors (IC50 = 0.5-3.9
You Zhou et al.
Molecules (Basel, Switzerland), 24(23) (2019-11-24)
As there are increased levels and activity of butyrylcholiesterase (BChE) in the late stage of Alzheimer's disease (AD), development of selective BChE inhibitors is of vital importance. In this study, a workflow combining computational technologies and biological assays were implemented
Gabriela Fernandes et al.
The journal of adhesive dentistry, 22(3), 265-274 (2020-05-22)
To investigate whether dental adhesives modified with polyacrylic acid copper iodide particles could inhibit esterase activity in vitro and the copper release rate from resin matrices, as well as the correlation between the two variables. Different concentrations of copper iodide
Cristobal Narvaez et al.
Chemosphere, 135, 75-82 (2015-04-29)
Inhibition of blood esterase activities by organophosphate (OP) pesticides has been used as a sensitive biomarker in birds. Furthermore, compared to mammalian vertebrates, less is known about the role of these enzyme activities in the digestive tracts of non-mammalian vertebrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service