Skip to Content
Merck
All Photos(1)

Key Documents

134996

Sigma-Aldrich

3-Iodobenzylamine hydrochloride

97%

Synonym(s):

(3-Iodophenyl)methanamine hydrochloride, m-Iodobenzylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$428.00

$428.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$428.00

About This Item

Linear Formula:
IC6H4CH2NH2 · HCl
CAS Number:
Molecular Weight:
269.51
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

$428.00


Estimated to ship onMarch 31, 2025


Request a Bulk Order

Quality Level

Assay

97%

mp

188-190 °C (lit.)

SMILES string

Cl.NCc1cccc(I)c1

InChI

1S/C7H8IN.ClH/c8-7-3-1-2-6(4-7)5-9;/h1-4H,5,9H2;1H

InChI key

PYFDZOCGFHIRST-UHFFFAOYSA-N

Application

3-Iodobenzylamine hydrochloride was used as the starting reagent in the synthesis of N6-(3-iodobenzyl)-2-substituted-adenosine derivatives[1]. It was used in the synthesis of 3′-C-methyl adenosine N6-substituted and N6/C-2 disubstituted derivatives and novel 2′-C-methyl analogues[2].

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H O Kim et al.
Journal of medicinal chemistry, 37(21), 3614-3621 (1994-10-14)
Adenosine derivatives bearing an N6-(3-iodobenzyl) group, reported to enhance the affinity of adenosine-5'-uronamide analogues as agonists at A3 adenosine receptors (J. Med. Chem. 1994, 37, 636-646), were synthesized starting from methyl beta-D-ribofuranoside in 10 steps. Binding affinities at A1 and
Loredana Cappellacci et al.
Journal of medicinal chemistry, 48(5), 1550-1562 (2005-03-04)
A number of 3'-C-methyl analogues of selective adenosine receptor agonists such as CPA, CHA, CCPA, 2'-Me-CCPA, NECA, and IB-MECA was synthesized to further investigate the subdomain of the receptor that binds the ribose moiety of the ligands. Affinity data at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service