Skip to Content
Merck
All Photos(1)

Key Documents

132497

Sigma-Aldrich

2,6-Dichloropyrazine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2Cl2N2
CAS Number:
Molecular Weight:
148.98
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39162101
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

55-58 °C (lit.)

functional group

chloro

SMILES string

Clc1cncc(Cl)n1

InChI

1S/C4H2Cl2N2/c5-3-1-7-2-4(6)8-3/h1-2H

InChI key

LSEAAPGIZCDEEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ling-Wei Kong et al.
Dalton transactions (Cambridge, England : 2003), 41(18), 5625-5633 (2012-03-17)
Cyclooligomerization of 2,6-dichloropyrazine 4 and benzyl 2,3-dihydroxybenzoate 5 under microwave irradiation resulted in a racemic pair of ester functionalized ortho-linked oxacalix[2]benzene[2]pyrazine 6, which was further transformed to the corresponding racemic carboxylic acid functionalized ortho-linked oxacalix[2]benzene[2]pyrazine 3. Both enantiomers of 3
Laixing Hu et al.
Bioorganic & medicinal chemistry, 21(21), 6732-6741 (2013-09-10)
Dicationic 2,6-diphenylpyrazines, aza-analogues and prodrugs were synthesized; evaluated for DNA affinity, activity against Trypanosoma brucei rhodesiense (T. b. r.) and Plasmodium falciparum (P. f.) in vitro, efficacy in T. b. r. STIB900 acute and T. b. brucei GVR35 CNS mouse
Benjamin J Coe et al.
The Journal of organic chemistry, 75(24), 8550-8563 (2010-11-18)
Six new dicationic 2D nonlinear optical (NLO) chromophores with pyrazinyl-pyridinium electron acceptors have been synthesized by nucleophilic substitutions of 2,6-dichloropyrazine with pyridyl derivatives. These compounds have been characterized as their PF(6)(-) salts by using various techniques including electronic absorption spectroscopy
Jeffrey L Katz et al.
Organic letters, 8(13), 2755-2758 (2006-06-16)
[structure: see text] Oxacalix[2]arene[2]hetarenes are formed in a single step by cyclooligomerization of meta-diphenols with meta-dichlorinated azaheterocycles. The high selectivity for cyclic tetramer formation results from thermodynamic product control. Macrocycles as large as oxacalix[5]arene[5]hetarenes have been isolated under nonequilibrating conditions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service