Skip to Content
Merck
All Photos(1)

Key Documents

130869

Sigma-Aldrich

1-Ethynylcyclopentanol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$84.00

$84.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$84.00

About This Item

Linear Formula:
HC≡CC5H8OH
CAS Number:
Molecular Weight:
110.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$84.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.474 (lit.)

bp

156-159 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

OC1(CCCC1)C#C

InChI

1S/C7H10O/c1-2-7(8)5-3-4-6-7/h1,8H,3-6H2

InChI key

LQMDOONLLAJAPZ-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Igor O Koshevoy et al.
Journal of the American Chemical Society, 134(15), 6564-6567 (2012-04-04)
We report a homoleptic Au-Cu alkynyl cluster that represents an unexplored class of luminescent materials with stimuli-responsive photophysical properties. The bimetallic complex formulated as [Au(2)Cu(2)(C(2)OHC(5)H(8))(4)](n) efficiently self-assembles from Au(SC(4)H(8))Cl, Cu(NCMe)(4)PF(6), and 1-ethynylcyclopentanol in the presence of NEt(3). This compound shows
Yahya H Dallal Bashi et al.
International journal of pharmaceutics, 569, 118574-118574 (2019-07-29)
With a dapivirine-releasing vaginal ring having successfully completed late-stage clinical testing for HIV prevention and currently undergoing regulatory review, there is now growing interest in next-generation multipurpose prevention technologies that seek to combine antiretroviral and contraceptive drugs within a single

Articles

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service