Skip to Content
Merck
4-Nitrobenzaldehyde 98% (GC)

4-Nitrobenzaldehyde

4-Nitrobenzaldehyde 98% (GC)
Synonym(s):
4-Nitrobenzaldehyde
Linear Formula:
4-(NO2)C6H4CHO
CAS Number:
Molecular Weight:
151.12

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing