Skip to Content
Merck
All Photos(3)

Key Documents

126896

Sigma-Aldrich

3-Fluorobenzylamine

97%

Synonym(s):

(3-Fluorophenyl)methanamine, 1-(3-Fluorophenyl)methanamine, 3-Fluorobenzenemethanamine, 3-Fluorophenylmethylamine, m-Fluorobenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2NH2
CAS Number:
Molecular Weight:
125.14
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39040408
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

liquid

refractive index

n20/D 1.514 (lit.)

density

1.097 g/mL at 25 °C (lit.)

functional group

amine
fluoro

SMILES string

NCc1cccc(F)c1

InChI

1S/C7H8FN/c8-7-3-1-2-6(4-7)5-9/h1-4H,5,9H2

InChI key

QVSVMNXRLWSNGS-UHFFFAOYSA-N

Application

3-Fluorobenzylamine has been used to study the rate of reaction of benzylamines with 1-Chloro-2,4-dinitrobenzene and toluene-p-sulphonyl chloride[1]. It has also been used in the synthesis of substituted amino-sulfonamide protease inhibitors (PIs) DPC 681 and DPC 684[2].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fischer A, et al.
Journal of the Chemical Society B: Physical Organic, 466-468 (1966)
R F Kaltenbach et al.
Antimicrobial agents and chemotherapy, 45(11), 3021-3028 (2001-10-16)
Human immunodeficiency virus (HIV) protease inhibitors (PIs) are important components of many highly active antiretroviral therapy regimens. However, development of phenotypic and/or genotypic resistance can occur, including cross-resistance to other PIs. Development of resistance takes place because trough levels of
A Paden King et al.
Dalton transactions (Cambridge, England : 2003), 48(18), 5987-6002 (2019-01-24)
Four cobalt(iii) complexes of the general formula [Co(Schiff base)(L)2]+, where L is ammonia (NH3) or 3-fluorobenzylamine (3F-BnNH2), were synthesized. The complexes were characterized by NMR spectroscopy, mass spectrometry, and X-ray crystallography. Their electrochemical properties, ligand substitution mechanisms, and ligand exchange
Hua-Li Yang et al.
European journal of medicinal chemistry, 138, 715-728 (2017-07-21)
A series of coumarin-pargyline hybrids (4a-x) have been designed, synthesized and evaluated as novel dual inhibitors of Alzheimer's disease (AD). Most of the compounds exhibited a potent ability to inhibit amyloid-β (Aβ) aggregation and monoamine oxidases. In particular, compound 4x
Simon H Rüdisser et al.
Journal of biomolecular NMR, 74(10-11), 579-594 (2020-06-20)
Fluorine NMR has recently gained high popularity in drug discovery as it allows efficient and sensitive screening of large numbers of ligands. However, the positive hits found in screening must subsequently be ranked according to their affinity in order to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service