Skip to Content
Merck
All Photos(2)

Key Documents

126551

Sigma-Aldrich

2,4-Dimethyl-1,3-pentadiene

98%

Synonym(s):

1,1,3-Trimethylbutadiene, 2,4-Dimethylpenta-2,4-diene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHC(CH3)=CH2
CAS Number:
Molecular Weight:
96.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

67 mmHg ( 37.7 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

94 °C (lit.)

density

0.744 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C\C(C)=C

InChI

1S/C7H12/c1-6(2)5-7(3)4/h5H,1H2,2-4H3

InChI key

CMSUNVGIWAFNBG-UHFFFAOYSA-N

Application

2,4-Dimethyl-1,3-pentadiene has been used to study the structure of its various conformational isomers and their vibrational spectra.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Zhao et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56(4), 671-679 (2000-05-04)
Ab initio was used to study the structure of various conformational isomers and their vibrational spectra of 2,4-dimethyl-1,3-pentadiene (2,4-PD) in detail. Two stable conformations, s-trans and s-cis, were found in which s-trans is more stable. The geometry of stable conformations
Damir Barisic et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(18), 4821-4832 (2019-02-05)
Targeting the synthesis of rare-earth-metal pentadienyl half-sandwich tetramethylaluminate complexes, homoleptic [Ln(AlMe4 )3 ] (Ln=Y, La, Ce, Pr, Nd, Lu) were treated with equimolar amounts of the potassium salts K(2,4-dmp) (2,4-dmp=2,4-dimethylpentadienyl), K(2,4-dipp) (2,4-dipp=2,4-diisopropylpentadienyl), and K(2,4-dtbp) (2,4-dtbp=2,4-di-tert-butylpentadienyl). The reactions involving the larger

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service