Skip to Content
Merck
All Photos(1)

Key Documents

511439

Sigma-Aldrich

trans-4-(Dimethylamino)-3-buten-2-one

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$130.00
25 G
$621.00

$130.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$130.00
25 G
$621.00

About This Item

Linear Formula:
(CH3)2NCH=CHCOCH3
CAS Number:
Molecular Weight:
113.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$130.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

97%

refractive index

n20/D 1.557 (lit.)

bp

101-102 °C/5 mmHg (lit.)

density

0.973 g/mL at 25 °C (lit.)

functional group

amine
ketone

storage temp.

2-8°C

SMILES string

[H]\C(=C(\[H])C(C)=O)N(C)C

InChI

1S/C6H11NO/c1-6(8)4-5-7(2)3/h4-5H,1-3H3/b5-4+

InChI key

QPWSKIGAQZAJKS-SNAWJCMRSA-N

General description

trans-4-(Dimethylamino)-3-buten-2-one is an α,β-unsaturated ketone. Its enthalpy of vaporization at boiling point (474.25K) is 41.416kjoule/mol.[1]

Application

trans-4-(Dimethylamino)-3-buten-2-one may be used in the preparation of 1,3,5-triacetylbenzene.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

225.0 °F - closed cup

Flash Point(C)

107.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 528-528 (2014)
1,3,5-Triacetylbenzene.
Alaimo PJ, et al.
Organic Syntheses, 87, 192-200 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service