Skip to Content
Merck
All Photos(1)

Documents

11270

Sigma-Aldrich

DL-Aspartic acid magnesium salt tetrahydrate

≥98.0%

Synonym(s):

Magnesium DL-aspartate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOOCCH(NH2)CH2COO]2Mg · 4H2O
CAS Number:
Molecular Weight:
360.56
Beilstein:
6029702
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

product name

DL-Aspartic acid magnesium salt tetrahydrate, ≥98.0%

Assay

≥98.0%

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

white

application(s)

peptide synthesis

SMILES string

OC(CC(N)C([O-])=O)=O.[Mg+2].C

InChI

1S/C4H7NO4.CH4.Mg/c5-2(4(8)9)1-3(6)7;;/h2H,1,5H2,(H,6,7)(H,8,9);1H4;/q;;+2/p-1

InChI key

YKAKEEZZQUOYRV-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service