Skip to Content
Merck
All Photos(3)

Documents

112127

Sigma-Aldrich

Diphenylacetonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CHCN
CAS Number:
Molecular Weight:
193.24
Beilstein:
1911160
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

181 °C/12 mmHg (lit.)

mp

71-73 °C (lit.)

SMILES string

N#CC(c1ccccc1)c2ccccc2

InChI

1S/C14H11N/c15-11-14(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,14H

InChI key

NEBPTMCRLHKPOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diphenylacetonitrile can be prepared in good yield by the dehydration of the amide of diphenyl acetic acid with phosphorous oxychloride.

Application

Diphenylacetonitrile undergoes anhydrous condensation with ethyl-4-bromo-butyrate. This is followed by its hydrolysis in alkaline medium that is used in the quantitative determination of 3-cyano-3,3-diphenylpropionic acid.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

245.5 °F - Pensky-Martens closed cup

Flash Point(C)

118.6 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M De Baere et al.
Journal of analytical toxicology, 20(3), 159-164 (1996-05-01)
A sensitive gas chromatographic procedure with nitrogen-phosphorus detection was developed for the quantitative determination of 3-cyano-3,3-diphenylpropionic acid, the acidic metabolite of the narcotic analgesic bezitramide (Burgodin). Gas chromatography with mass spectrometric detection was used for confirmation. An internal standard, 5-cyano-5,5-diphenylvaleric
Preparation of diphenylacetonitrile.
W B REID et al.
Journal of the American Chemical Society, 70(10), 3515-3515 (1948-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service