Skip to Content
Merck
All Photos(1)

Key Documents

10987

Sigma-Aldrich

Tetrakis(triphenylphosphine)palladium, polymer-bound

extent of labeling: ~0.06 mmol/g loading

Synonym(s):

Palladium-tetrakis(triphenylphosphine), polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reaction type: solution phase peptide synthesis
reagent type: catalyst

extent of labeling

~0.06 mmol/g loading

matrix

crosslinked with 1% DVB

functional group

phosphine

storage temp.

2-8°C

SMILES string

[Pd].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9.c%10ccc(cc%10)P(c%11ccccc%11)c%12ccccc%12

InChI

1S/4C18H15P.Pd/c4*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h4*1-15H;

InChI key

NFHFRUOZVGFOOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Polymer bound palladium catalyst

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I. Fenger et al.
Tetrahedron Letters, 39, 4287-4287 (1998)
Mira Okshevsky et al.
MicrobiologyOpen, 7(2), e00552-e00552 (2017-11-23)
Bacillus cereus is one of the most common opportunistic pathogens causing foodborne illness, as well as a common source of contamination in the dairy industry. B. cereus can form robust biofilms on food processing surfaces, resulting in food contamination due to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service