Skip to Content
Merck
All Photos(1)

Documents

104361

Sigma-Aldrich

Tetrafluorohydroquinone

95%

Synonym(s):

2,3,5,6-Tetrafluorobenzene-1,4-diol, 2,3,5,6-Tetrafluorohydroquinone, Tetrafluoro-1,4-dihydroquinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F4-1,4-(OH)2
CAS Number:
Molecular Weight:
182.07
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

mp

172-174 °C (lit.)

SMILES string

Oc1c(F)c(F)c(O)c(F)c1F

InChI

1S/C6H2F4O2/c7-1-2(8)6(12)4(10)3(9)5(1)11/h11-12H

InChI key

ZSDAMBJDFDRLSS-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Mao et al.
Free radical biology & medicine, 147, 150-158 (2019-12-21)
Penicillamine has been widely-used clinically as a copper-chelating drug for the treatment of copper-overload in Wilson's disease. In this study, we found that penicillamine provided marked protection against cytotoxicity induced by tetrachlorohydroquinone (TCHQ), a major toxic metabolite of the well-known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service