Skip to Content
Merck
All Photos(2)

Key Documents

101087

Sigma-Aldrich

2-Bromo-4′-phenylacetophenone

technical grade

Synonym(s):

ω-Bromo-4-phenylacetophenone, 4-Phenylphenacyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4COCH2Br
CAS Number:
Molecular Weight:
275.14
Beilstein:
513838
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

mp

123-125 °C (lit.)

functional group

bromo
ketone
phenyl

SMILES string

BrCC(=O)c1ccc(cc1)-c2ccccc2

InChI

1S/C14H11BrO/c15-10-14(16)13-8-6-12(7-9-13)11-4-2-1-3-5-11/h1-9H,10H2

InChI key

KGHGZRVXCKCJGX-UHFFFAOYSA-N

Gene Information

human ... PTPN6(5777)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Caproşu et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 93(1), 175-177 (1989-01-01)
This time the synthesis of some new derivatives obtained by the quaternary substitution of one of the heterocyclic nitrogen atoms and by reactions of dipolar cycloaddition is presented. The results of the tests on their antimicrobial and antiinflammatory actions are
Sedigheh Abedanzadeh et al.
Dalton transactions (Cambridge, England : 2003), 49(42), 14891-14907 (2020-10-20)
A rational approach was adopted to design high-potential metal-based antitumor agents. A series of organometallic Pd(ii) complexes with a general formula of [Pd{κ2(C,C)-[(C6H4-2)PPh2]CH(CO)C6H4Ph-4}{κ2(N,O)}] (N,O = alanine (Pd-A), valine (Pd-V), leucine (Pd-L), l-isoleucine (Pd-I) and phenylalanine (Pd-F)) were prepared by cyclopalladation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service