Skip to Content
Merck
All Photos(2)

Key Documents

CLS430909

Corning® microcentrifuge tubes with screw cap

1.5 mL microcentrifuge tube, polypropylene, w/ attached screw cap, w/ 0-ring, sterile, natural, 500/cs

Synonym(s):

centrifuge tubes, micrcentrifuge tubes, microfuge tubes, screw cap microcentrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
eCl@ss:
32119210
NACRES:
NB.11

material

conical bottom tube
natural polypropylene tube
screw top cap

description

O-ring (black silicone, rubber O-ring in cap)

sterility

sterile

feature

self-standing: no conical bottom

packaging

pack of 50
case of 500

manufacturer/tradename

Corning 430909

parameter

20000 × g max. RCF

cap diam.

12.7 mm

capacity

1.5 mL

tube O.D.

10.7 mm

color

natural

fitting

cap (screw cap)

Looking for similar products? Visit Product Comparison Guide

General description

  • 1.5 mL polypropylene microcentrifuge tubes feature screw caps that provide a tight secure seal
  • Attached cap with silicone O-ring
  • Withstands a maximum RCF of 20,000xg
  • Sterile
Corning polypropylene microcentrifuge tubes feature screw caps that provide a tight, secure seal.
  • Maximum RCF is 20,000
  • No marking spot

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Reverse transcription (RT) is the process of converting RNA to cDNA using a reverse transcriptase enzyme and dNTPs.

Hot Start dNTPs block DNA polymerase until heat activation, enhancing PCR specificity.

Primer Concentration Optimization Protocol creates reaction matrix for testing primer concentrations against various partners.

Reverse transcription analyzes mRNA gene expression, facing challenges like transcript half-life differences and temporal patterns.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service