Skip to Content
Merck
All Photos(1)

Key Documents

G2417

Supelco

Gabexate mesylate

analytical standard, for drug analysis

Synonym(s):

4-[[6-[(Aminoiminomethyl)amino]-1-oxohexyl]oxyl]benzoic acid ethyl ester mesylate salt, FOY, Megacart

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23N3O4 · CH4O3S
CAS Number:
Molecular Weight:
417.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CS(O)(=O)=O.CCOC(=O)c1ccc(OC(=O)CCCCCNC(N)=N)cc1

InChI

1S/C16H23N3O4.CH4O3S/c1-2-22-15(21)12-7-9-13(10-8-12)23-14(20)6-4-3-5-11-19-16(17)18;1-5(2,3)4/h7-10H,2-6,11H2,1H3,(H4,17,18,19);1H3,(H,2,3,4)

InChI key

DNTNDFLIKUKKOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gabexate mesylate is a non-antigenic synthetic inhibitor of trypsin-like serine proteinases, which finds therapeutic applications in the treatment of disseminated intravascular coagulation and pancreatitis. It also plays the role of an anticoagulant in hemodialysis therapy.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoki Nishiyama et al.
The American journal of emergency medicine, 30(7), 1219-1223 (2011-12-30)
We hypothesized that antithrombin is more effective for disseminated intravascular coagulation (DIC) than is gabexate mesilate, which is a protease inhibitor, suggested from the previous studies. Initially, we compared the effects of antithrombin and gabexate mesilate for treating infection-related DIC.
S Aosasa et al.
Shock (Augusta, Ga.), 15(2), 101-105 (2001-02-28)
If the inflammatory response becomes excessive or uncontrolled by some stimuli, inappropriate inflammatory responses occur. Monocytes are extremely important cells for regulating the cytokine network and tumor necrosis factor alpha (TNFalpha) and interleukin- (IL) 10, which are mainly synthesized by
Maria Dorly Del Curto et al.
Journal of pharmaceutical sciences, 100(8), 3251-3259 (2011-04-06)
It is well known that the intestinal stability and absorption of protein drugs are improved when enzyme inhibitors/permeation enhancers are coadministered. Recently, it was hypothesized that an increased effectiveness of these adjuvants might be achieved by timing their release prior
Kohei Uchimura et al.
American journal of physiology. Renal physiology, 303(7), F939-F943 (2012-07-27)
Aldosterone plays an important role in the regulation of blood pressure by modulating the activity of the epithelial sodium channel (ENaC) that consists of α-, β-, and γ-subunits. Aldosterone induces a molecular weight shift of γENaC from 85 to 70
James A Reihill et al.
Biomolecules, 10(4) (2020-04-02)
Epithelial barrier dysfunction, characteristic of allergic airway disease may be, at least in part, due to the action of allergen-associated protease activities. Cockroach allergy is a major global health issue, with cockroaches containing considerable serine trypsin-like protease (TLP) activity. The

Articles

Analytical Enzyme Chymotrypsin: Chymotrypsin is produced in the acinar cells of the pancreas as the inactive precursor, chymotrypsinogen.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service