Skip to Content
Merck
All Photos(1)

Key Documents

SML3210

Sigma-Aldrich

Sotrastaurin

≥98% (HPLC)

Synonym(s):

3-(1H-Indol-3-yl)-4-(2-(4-methylpiperazin-1-yl)quinazolin-4-yl)-1H-pyrrole-2,5-dione, 3-(1H-Indol-3-yl)-4-[2-(4-methyl-piperazin-1-yl)quinazolin-4-yl]pyrrole-2,5-dione, AEB 071, AEB-071, AEB071, NVP-AEB071, Sotrastaurin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$112.00
25 MG
$453.00

$112.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$112.00
25 MG
$453.00

About This Item

Empirical Formula (Hill Notation):
C25H22N6O2
CAS Number:
Molecular Weight:
438.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$112.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

faint brown to dark red

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(NC1=O)C(C2=CNC3=C2C=CC=C3)=C1C4=C5C=CC=CC5=NC(N6CCN(CC6)C)=N4

InChI

1S/C25H22N6O2/c1-30-10-12-31(13-11-30)25-27-19-9-5-3-7-16(19)22(28-25)21-20(23(32)29-24(21)33)17-14-26-18-8-4-2-6-15(17)18/h2-9,14,26H,10-13H2,1H3,(H,29,32,33)

InChI key

OAVGBZOFDPFGPJ-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, ATP site-targeting, potent and selective protein kinase C (PKC) inhibitor with in vivo immunosuppressive efficacy.
Sotrastaurin (AEB071) is an orally active, ATP site-targeting, potent and selective protein kinase C (PKC) inhibitor (Ki in nM/isoform = 0.95/α, 0.64/βI, 2.1/δ, 3.2/ε, 1.8/η and 2.2/θ; IC50 = 2.1/α, 2.0/βI, 1.3/δ, 6.2/ε, 6.1/η and 1.0/θ with 10 μM ATP). Sotrastaurin inhibits TCR/CD28-mediated T cell activation in cultures (IC50 = 54 nM by Jurkat IL-2 promoter reporter line) and exhibits in vivo immunosuppressive efficacy in rat models of local graft versus host (GvH) reaction and cardiac transplantation (10 and 30 mg/kg p.o. q.d. or b.i.d.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service