Skip to Content
Merck
All Photos(2)

Key Documents

308315

Sigma-Aldrich

Lithium hexafluoroarsenate(V)

greener alternative

98%

Synonym(s):

Hexafluoroarsenate(V) lithium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiAsF6
CAS Number:
Molecular Weight:
195.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
powder
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

powder

reaction suitability

reagent type: catalyst
core: arsenic

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].F[As-](F)(F)(F)(F)F

InChI

1S/AsF6.Li/c2-1(3,4,5,6)7;/q-1;+1

InChI key

GTZQZEYBOGZTEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium hexafluoroarsenate(V) is a class of electrolytic materials that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.[1][2][3][4]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

The surface chemistry of Li electrodes in ethereal LiAsF6 was tested with new salts for potential battery systems.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling-Yun Yang et al.
Angewandte Chemie (International ed. in English), 53(14), 3631-3635 (2014-03-07)
A new category of crystalline polymer electrolyte prepared by the supramolecular self-assembly of polyethylene oxide (PEO), α-cyclodextrin (α-CD), and LiAsF6 is reported. The polymer electrolyte consists of the nanochannels formed by α-CDs in which the PEO/Li(+) complexes are confined. The
The Li-ion rechargeable battery: a perspective
Goodenough JB and Park K
Journal of the American Chemical Society, 135(4), 1167-1176 (2013)
Challenges for rechargeable Li batteries
Goodenough JB and Kim Y
Chemistry of Materials, 22(3), 587-603 (2009)
Electrodes with high power and high capacity for rechargeable lithium batteries
Kang K, et al.
Science, 311(5763), 977-980 (2006)
Towards greener and more sustainable batteries for electrical energy storage
Larcher D and Tarascon J
Nature Chemistry, 7(1), 19-19 (2015)

Articles

Nanomaterials for Energy Storage in Lithium-ion Battery Applications

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

Solid-state Li batteries: Review of solid electrolytes, ion conduction, structures, and electrochemical processes.

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service