Skip to Content
Merck
Lithium hexafluoroarsenate(V) 99.9% trace metals basis

Lithium hexafluoroarsenate(V)

Lithium hexafluoroarsenate(V) 99.9% trace metals basis
Synonym(s):
Hexafluoroarsenate(V) lithium
Linear Formula:
LiAsF6
CAS Number:
Molecular Weight:
195.85

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing