Merck
  • An optimized method to process mouse CNS to simultaneously analyze neural cells and leukocytes by flow cytometry.
An optimized method to process mouse CNS to simultaneously analyze neural cells and leukocytes by flow cytometry.
Journal of neuroscience methods (2015-03-31)
Laurine Legroux, Camille L Pittet, Diane Beauseigle, Gabrielle Deblois, Alexandre Prat, Nathalie Arbour
ABSTRACT
Flow cytometry is an efficient and powerful technique to characterize and quantify numerous cells. However, the strengths of this technique have not been widely harnessed in neurosciences due to the critical step of CNS tissue preparation into a single cell suspension. Previous reports assessed either neural cells or infiltrating leukocytes but simultaneous detection has not been extensively implemented. We optimized CNS tissue preparation for flow cytometry analysis. We subjected CNS tissue from individual adult mice to different digestion protocols and Percoll™ methods. We quantified and characterized by flow cytometry neural cells (neurons, oligodendrocytes, microglia) and leukocytes (macrophages, T lymphocytes). The one step Percoll™ method significantly increased cell yield compared to the gradient Percoll™ method. The collagenase D+DNase I digestion led to the maximal cell number recovery while preserving cell marker (O4, NeuN, CD45, CD11b, CD3, CD4, CD8) integrity compared to papain, trypsin digestion, and no digestion. The combination of collagenase D+DNase I digestion and one step Percoll™ method was optimal for the recovery and analysis of cells from the CNS of naïve and experimental autoimmune encephalomyelitis (multiple sclerosis model) mice. Although flow cytometry does not reveal CNS localization, this technique allows concurrent quantification of multiple parameters. In contrast to other protocols, our novel method simultaneously analyzes neural and immune cells in individual mice in healthy and pathological conditions. We strongly believe that the field of neurosciences will benefit from an optimal use of flow cytometry to elucidate physiological and pathological processes.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Screen cup for CD-1, size 85 mL
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Roche
DNase I, from bovine pancreas
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT)
Sigma-Aldrich
L-Cysteine hydrochloride hydrate, 99%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Screens for CD-1, size 100 mesh
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Saponin from quillaja bark, Sapogenin content ≥10 %
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.5-101.0%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
L-Cysteine hydrochloride monohydrate, from non-animal source, suitable for cell culture, meets EP, USP testing specifications
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES