Merck
  • Methods for differential and quantitative analyses of brain neurosteroid levels by LC/MS/MS with ESI-enhancing and isotope-coded derivatization.
Methods for differential and quantitative analyses of brain neurosteroid levels by LC/MS/MS with ESI-enhancing and isotope-coded derivatization.
Journal of pharmaceutical and biomedical analysis (2015-09-12)
Tatsuya Higashi, Naoto Aiba, Tomoya Tanaka, Kazumi Yoshizawa, Shoujiro Ogawa
ABSTRACT
The analysis of changes in the brain neurosteroid (NS) levels due to various stimuli can contribute to the elucidation of their physiological roles, and the discovery and development of new antipsychotic agents targeting neurosteroidogenesis. We developed methods for the differential and quantitative analyses of the brain levels of allopregnanolene (AP) and its precursor, pregnenolone (PREG), using liquid chromatography/electrospray ionization-tandem mass spectrometry (LC/ESI-MS/MS) combined with derivatization using 2-hydrazino-1-methylpyridine (HMP) and its isotope-coded analogue, (2)H3-HMP (d-HMP). For the differential analysis, the brain sample of an untreated rat was derivatized with HMP, while the brain sample of a treated (stressed or drug-administered) rat was derivatized with d-HMP. The two derivatives were mixed and then subjected to LC/ESI-MS/MS. The stress- and drug (clozapine and fluoxetine)-evoked increases in the brain AP and PREG levels were accurately analyzed by the developed method. It was also possible to determine the absolute concentrations of the brain steroids when a deuterium-coded moiety was introduced to the standard steroids of known amounts by the derivatization and the resulting derivatives were used as internal standards. The HMP-derivatization enabled the highly sensitive detection and the use of d-HMP significantly improved the assay precision [the intra- (n=5) and inter-assay (n=5) relative standard deviations did not exceed 13.7%] and accuracy (analytical recovery ranged from 98.7 to 106.7%).
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
3,4-Dihydro-2H-pyran, 97%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Haloperidol, powder
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture