Skip to Content
Merck
All Photos(3)

Key Documents

H1512

Sigma-Aldrich

Haloperidol

≥98% (TLC), powder, dopamine receptor antagonist

Synonym(s):

4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone, 4-[4-(4-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone, 4-[4-(p-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23ClFNO2
CAS Number:
Molecular Weight:
375.86
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Haloperidol, powder

form

powder

Quality Level

color

white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.39 mg/mL
0.1 M HCl: 3 mg/mL
DMSO: soluble
H2O: insoluble
ethanol: soluble

originator

Johnson & Johnson

SMILES string

OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3ccc(Cl)cc3

InChI

1S/C21H23ClFNO2/c22-18-7-5-17(6-8-18)21(26)11-14-24(15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,26H,1-2,11-15H2

InChI key

LNEPOXFFQSENCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Haloperidol has been used:
  • in ethanol to serves as an inhibitor of Erg2p
  • to address the mechanism of haloperidol in ferroptosis using hepatocellular carcinoma cells: Hep G2 and Huh-7 cell lines
  • in receptor internalization assay
  • as an antipsychotic drug in Dulbecco′s Modified Eagle medium

Biochem/physiol Actions

Haloperidol is a butyrophenone antipsychotic. It is also classified as a neuroleptic (powerful tranquilizer). Haloperidol acts as a D2, D3, and D4 dopamine receptor antagonist and thus causes Parkinson′s disorder. It also has a negative effect on the central nervous system.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CRC Desk Reference of Clinical Pharmacology null
Haloperidol changes mRNA expression of a QKI splice variant in human astrocytoma cells
Jiang L, et al.
BioMed Central Pharmacology, 9(1), 6-6 (2009)
Cdk5 phosphorylates dopamine D2 receptor and attenuates downstream signaling
Jeong J, et al.
Testing, 8(12), e84482-e84482 (2013)
Haloperidol, a sigma receptor 1 antagonist, promotes ferroptosis in hepatocellular carcinoma cells
Bai T, et al.
Biochemical and Biophysical Research Communications, 491(4), 919-925 (2017)
Yeast sphingolipids do not need to contain very long chain fatty acids
Cerantola V, et al.
The Biochemical Journal, 401(1), 205-216 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service